Assay Details
Target Gene Details
Entrez Gene ID: | 5024 |
Gene Name: | purinergic receptor P2X 3 |
Gene Aliases: |
P2X3 |
Location: |
Chr.11:57335943-57372399 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX3 | NM_002559.3 | 8 | 982 | NP_002550.2 |
| XM_011545069.2 | 9 | 1643 | XP_011543371.1 | |
| XM_011545070.2 | 9 | 949 | XP_011543372.1 | |
| XM_011545071.2 | 8 | 1511 | XP_011543373.1 | |
| XM_011545072.2 | 9 | 1775 | XP_011543374.1 | |
| XM_011545073.1 | 6 | 499 | XP_011543375.1 | |
| AB016608.1 | 8 | 798 | BAA76515.1 | |
| BC069162.1 | 8 | 874 | AAH69162.1 | |
| BC074792.2 | 8 | 829 | AAH74792.1 | |
| BC074793.2 | 8 | 829 | AAH74793.1 | |
| BC109287.2 | 8 | 832 | AAI09288.1 |
More Information
Set Membership: |
|

Genomic Map