Assay Details
Target Gene Details
Entrez Gene ID: | 5101 |
Gene Name: | protocadherin 9 |
Gene Aliases: |
- |
Location: |
Chr.13:66302834-67230336 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH9 | NM_001318372.1 | 5 | 5468 | NP_001305301.1 |
| NM_001318373.1 | 4 | 5366 | NP_001305302.1 | |
| NM_020403.4 | 4 | 5492 | NP_065136.1 | |
| NM_203487.2 | 5 | 5594 | NP_982354.1 | |
| XM_017020619.1 | 5 | 5362 | XP_016876108.1 | |
| AF169692.2 | 3 | 4918 | AAF89689.2 | |
| AL834258.1 | 4 | 4445 | CAD38933.1 | |
| BC136626.1 | 5 | 5076 | ||
| BC136627.1 | 5 | 5076 | ||
| BC144321.1 | 5 | 4950 | ||
| BX537422.1 | 4 | 5490 | CAD97664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043603 | Chr.13:65487421 - 66772963 on Build GRCh38 | Gain |
|
| esv3632502 | Chr.13:66253178 - 66349265 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map