Assay Details
Target Gene Details
Entrez Gene ID: | 115024 |
Gene Name: | 5'-nucleotidase, cytosolic IIIB |
Gene Aliases: |
NT5C3L, cN-IIIB |
Location: |
Chr.17:41825082-41836276 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C3B | NM_052935.4 | NP_443167.4 | ||
| NR_033464.1 | ||||
| NR_033465.1 | ||||
| XM_006721669.3 | XP_006721732.1 | |||
| XM_006721670.3 | XP_006721733.1 | |||
| XM_011524276.2 | XP_011522578.1 | |||
| XM_011524277.2 | XP_011522579.1 | |||
| XM_017024127.1 | XP_016879616.1 | |||
| BC013742.1 | AAH13742.2 | |||
| BC014132.2 | AAH14132.2 | |||
| BC016971.1 | AAH16971.2 | |||
| BC067788.1 | AAH67788.1 | |||
| BF339623.1 | ||||
| BX365588.2 | ||||
| BX460247.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3174n100 | Chr.17:41778258 - 41843183 on Build GRCh38 | Gain |
|
| nsv827994 | Chr.17:41823755 - 41836053 on Build GRCh38 | Gain |
|
| nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map