Assay Details
Target Gene Details
| Entrez Gene ID: | 25885 | 
| Gene Name: | RNA polymerase I subunit A | 
| Gene Aliases: | A190, AFDCIN, RPA1, RPA194, RPO1-4, RPO14 | 
| Location: | Chr.2:86026328-86106155 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 28 - Exon 29 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| POLR1A | NM_015425.4 | NP_056240.2 | ||
| AK025568.1 | BAB15171.1 | |||
| AK294447.1 | ||||
| AK302458.1 | ||||
| AK304131.1 | ||||
| BC017115.1 | ||||
| BC117173.1 | ||||
| BC126303.1 | ||||
| BC143345.1 | ||||
| U33460.1 | AAC99959.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2751903 | Chr.2:86026419 - 86278330 on Build GRCh38 | Gain |  SNORD94  REEP1  POLR1A  PTCD3  MRPL35  IMMT  MIR4779 | 
| esv32617 | Chr.2:86029518 - 86033162 on Build GRCh38 | Loss |  POLR1A | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top