Assay Details
Target Gene Details
| Entrez Gene ID: | 4595 | 
| Gene Name: | mutY DNA glycosylase | 
| Gene Aliases: | MYH | 
| Location: | Chr.1:45329242-45340925 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 9 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |  TOE1  MUTYH  TESK2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top