Assay Details
Target Gene Details
Entrez Gene ID: | 4199 |
Gene Name: | malic enzyme 1 |
Gene Aliases: |
HUMNDME, MES |
Location: |
Chr.6:83210389-83431219 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ME1 | NM_002395.5 | 14 | 1882 | NP_002386.1 |
| XM_011535836.2 | 13 | 1607 | XP_011534138.1 | |
| AK223417.1 | 14 | 1708 | BAD97137.1 | |
| AK289783.1 | 14 | 1710 | ||
| AK301875.1 | 12 | 1424 | ||
| AK302777.1 | 13 | 1607 | ||
| BC017403.1 | 5 | 602 | AAH17403.1 | |
| BC025246.1 | 14 | 1698 | AAH25246.1 | |
| L34035.1 | 14 | 1629 | AAB01380.1 | |
| U43944.1 | 14 | 1596 | AAC50613.1 | |
| X77244.1 | 14 | 1617 | CAA54460.1 |
More Information
Set Membership: |
|

Genomic Map