Assay Details
Target Gene Details
Entrez Gene ID: | 202374 |
Gene Name: | serine/threonine kinase 32A |
Gene Aliases: |
YANK1 |
Location: |
Chr.5:147234963-147401996 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK32A | NM_001112724.1 | NP_001106195.1 | ||
| NM_001287740.1 | NP_001274669.1 | |||
| NM_145001.3 | NP_659438.1 | |||
| XM_011537577.2 | XP_011535879.1 | |||
| XM_011537579.2 | XP_011535881.1 | |||
| XM_017009213.1 | XP_016864702.1 | |||
| AK094580.1 | ||||
| AK299099.1 | ||||
| AK315942.1 | ||||
| BC021666.1 | AAH21666.1 | |||
| DA110948.1 | ||||
| DR006392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599938 | Chr.5:146789948 - 147426802 on Build GRCh38 | Gain |
|
| nsv519847 | Chr.5:146786815 - 147433084 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map