Assay Details
Target Gene Details
Entrez Gene ID: | 3788 |
Gene Name: | potassium voltage-gated channel modifier subfamily S member 2 |
Gene Aliases: |
KV9.2 |
Location: |
Chr.8:98427022-98430797 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNS2 | NM_020697.3 | 2 | 2485 | NP_065748.1 |
| AB032970.1 | 2 | 2290 | ||
| BC034778.1 | 2 | 2485 | AAH34778.1 |
Target Gene Details
Entrez Gene ID: | 6788 |
Gene Name: | serine/threonine kinase 3 |
Gene Aliases: |
KRS1, MST2 |
Location: |
Chr.8:98429677-98944329 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK3 | XM_011517251.1 | 12 | 5037 | XP_011515553.1 |
| XM_011517253.2 | 10 | 4706 | XP_011515555.1 | |
| XM_011517255.2 | 8 | 5159 | XP_011515557.1 | |
| XM_011517258.1 | 7 | 4271 | XP_011515560.1 | |
| XM_017013759.1 | 14 | 5372 | XP_016869248.1 | |
| XM_017013760.1 | 13 | 5235 | XP_016869249.1 | |
| XM_017013763.1 | 8 | 4351 | XP_016869252.1 | |
| XM_017013764.1 | 7 | 5096 | XP_016869253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034492 | Chr.8:98424113 - 98444288 on Build GRCh38 | Gain |
|
| nsv831406 | Chr.8:98278517 - 98469848 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map