Assay Details
Target Gene Details
Entrez Gene ID: | 54946 |
Gene Name: | solute carrier family 41 member 3 |
Gene Aliases: |
SLC41A1-L2 |
Location: |
Chr.3:126006357-126101550 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009333 | Chr.3:126006809 - 126041785 on Build GRCh38 | Loss |
|
| nsv524971 | Chr.3:126024828 - 126155545 on Build GRCh38 | Loss |
|
| nsv591492 | Chr.3:126014238 - 126076708 on Build GRCh38 | Gain |
|
| nsv591465 | Chr.3:125283331 - 126140524 on Build GRCh38 | Gain |
|
| nsv829716 | Chr.3:125928159 - 126116976 on Build GRCh38 | Gain+Loss |
|
| nsv1011457 | Chr.3:125287676 - 126146923 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map