Assay Details
Target Gene Details
Entrez Gene ID: | 219 |
Gene Name: | aldehyde dehydrogenase 1 family member B1 |
Gene Aliases: |
ALDH5, ALDHX |
Location: |
Chr.9:38392664-38398665 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1B1 | NM_000692.4 | 2 | 964 | NP_000683.3 |
| XM_011517802.2 | 2 | 904 | XP_011516104.1 | |
| AK225950.1 | 2 | 877 | ||
| AK297020.1 | ||||
| AK313344.1 | 2 | 927 | ||
| BC001619.1 | 2 | 901 | AAH01619.1 | |
| BT007418.1 | 1 | 811 | AAP36086.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018488 | Chr.9:38021455 - 38455960 on Build GRCh38 | Gain |
|
| nsv8445 | Chr.9:38283618 - 38685720 on Build GRCh38 | Gain+Loss |
|
| esv34172 | Chr.9:38335478 - 38436995 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map