Assay Details
Target Gene Details
Entrez Gene ID: | 10219 |
Gene Name: | killer cell lectin like receptor G1 |
Gene Aliases: |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
Location: |
Chr.12:8989079-9255692 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRG1 | XM_017018685.1 | XP_016874174.1 |
Target Gene Details
Entrez Gene ID: | 5858 |
Gene Name: | PZP, alpha-2-macroglobulin like |
Gene Aliases: |
CPAMD6 |
Location: |
Chr.12:9140730-9208370 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PZP | NM_002864.2 | NP_002855.2 | ||
| XM_011520804.2 | XP_011519106.1 | |||
| XM_011520805.2 | XP_011519107.1 | |||
| XM_017019763.1 | XP_016875252.1 | |||
| XM_017019764.1 | XP_016875253.1 | |||
| XM_017019765.1 | XP_016875254.1 | |||
| XM_017019766.1 | XP_016875255.1 | |||
| AK313639.1 | ||||
| BC111756.1 | AAI11757.1 | |||
| X54380.1 | CAA38255.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523958 | Chr.12:9150844 - 9396476 on Build GRCh38 | Loss |
|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv527404 | Chr.12:9098005 - 9282648 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map