Assay Details
Target Gene Details
Entrez Gene ID: | 8382 |
Gene Name: | NME/NM23 family member 5 |
Gene Aliases: |
NM23-H5, NM23H5, RSPH23 |
Location: |
Chr.5:138115172-138139443 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME5 | NM_003551.2 | 6 | 691 | NP_003542.1 |
| XM_006714716.3 | 4 | 499 | XP_006714779.1 | |
| XM_017009945.1 | 5 | 1456 | XP_016865434.1 | |
| AF067724.1 | 6 | 661 | AAC64358.1 | |
| AK312181.1 | ||||
| BC026182.2 | 6 | 663 | AAH26182.1 | |
| BM979998.1 | 1 | 553 | ||
| KU178416.1 | ||||
| KU178417.1 | ||||
| U90450.1 | 6 | 637 | AAC69440.1 | |
| Y14992.1 | 5 | 627 | CAA75226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575884 | Chr.5:138058161 - 138118429 on Build GRCh38 | Gain |
|
| esv3606913 | Chr.5:137856857 - 138124533 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map