Assay Details
Target Gene Details
Entrez Gene ID: | 9201 |
Gene Name: | doublecortin like kinase 1 |
Gene Aliases: |
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK |
Location: |
Chr.13:35768652-36132020 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCLK1 | NM_001195415.1 | 14 | 4051 | NP_001182344.1 |
| NM_001195416.1 | 13 | 3977 | NP_001182345.1 | |
| NM_004734.4 | 18 | 4922 | NP_004725.1 | |
| XM_005266592.2 | 17 | 4801 | XP_005266649.1 | |
| XM_006719893.2 | 17 | 4835 | XP_006719956.1 | |
| XM_017020847.1 | 17 | 4812 | XP_016876336.1 | |
| AB002367.1 | 18 | 4851 | ||
| BC152456.1 | 18 | 4851 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832580 | Chr.13:35627110 - 35793577 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map