Assay Details
Target Gene Details
Entrez Gene ID: | 1809 |
Gene Name: | dihydropyrimidinase like 3 |
Gene Aliases: |
CRMP-4, CRMP4, DRP-3, DRP3, LCRMP, ULIP, ULIP-1 |
Location: |
Chr.5:147390808-147510056 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYSL3 | NM_001197294.1 | 14 | 3435 | NP_001184223.1 |
| NM_001387.2 | 14 | 3005 | NP_001378.1 | |
| XM_011537574.1 | 15 | 3389 | XP_011535876.1 | |
| BC077077.1 | 14 | 3435 | AAH77077.1 | |
| D78014.1 | 14 | 3005 | BAA11192.1 |
Target Gene Details
Entrez Gene ID: | 202374 |
Gene Name: | serine/threonine kinase 32A |
Gene Aliases: |
YANK1 |
Location: |
Chr.5:147234963-147401996 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK32A | XM_011537577.2 | XP_011535879.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599938 | Chr.5:146789948 - 147426802 on Build GRCh38 | Gain |
|
| nsv519847 | Chr.5:146786815 - 147433084 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76958182] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map