Assay Details
Target Gene Details
Entrez Gene ID: | 23704 |
Gene Name: | potassium voltage-gated channel subfamily E regulatory subunit 4 |
Gene Aliases: |
MIRP3 |
Location: |
Chr.2:223051930-223055637 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNE4 | NM_080671.3 | NP_542402.3 | ||
| AK294423.1 | ||||
| AK313203.1 | ||||
| AY065987.1 | AAL49979.1 | |||
| BC014429.1 | AAH14429.1 | |||
| BP348485.1 | ||||
| BP359441.1 | ||||
| BP362695.1 | ||||
| DQ192293.1 | ABB00306.1 |
More Information
Set Membership: |
|

Genomic Map