Assay Details
Target Gene Details
Entrez Gene ID: | 28991 |
Gene Name: | COMM domain containing 5 |
Gene Aliases: |
HCARG, HT002 |
Location: |
Chr.8:144837978-144853578 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD5 | AK023070.1 | BAB14389.1 |
Target Gene Details
Entrez Gene ID: | 7553 |
Gene Name: | zinc finger protein 7 |
Gene Aliases: |
HF.16, KOX4, zf30 |
Location: |
Chr.8:144827500-144848070 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF7 | NM_001282795.1 | 5 | 783 | NP_001269724.1 |
| NM_001282796.1 | NP_001269725.1 | |||
| NM_001282797.1 | 4 | 623 | NP_001269726.1 | |
| NM_003416.3 | 5 | 750 | NP_003407.1 | |
| XM_006716654.2 | 3 | 514 | XP_006716717.1 | |
| XM_006716656.3 | 3 | 503 | XP_006716719.1 | |
| XM_011517291.2 | 5 | 797 | XP_011515593.1 | |
| XM_011517292.2 | 5 | 1131 | XP_011515594.1 | |
| XM_011517293.2 | 5 | 1130 | XP_011515595.1 | |
| XM_011517294.2 | 5 | 909 | XP_011515596.1 | |
| XM_011517295.2 | 5 | 690 | XP_011515597.1 | |
| XM_011517296.2 | 5 | 701 | XP_011515598.1 | |
| XM_011517297.2 | 3 | 550 | XP_011515599.1 | |
| XM_017013813.1 | 5 | 794 | XP_016869302.1 | |
| XM_017013814.1 | 4 | 1096 | XP_016869303.1 | |
| XM_017013815.1 | 4 | 1093 | XP_016869304.1 | |
| XM_017013816.1 | XP_016869305.1 | |||
| XM_017013817.1 | XP_016869306.1 | |||
| AB209619.1 | 2 | 4139 | BAD92856.1 | |
| AK096025.1 | BAC04677.1 | |||
| AK300001.1 | 5 | 718 | ||
| AK301561.1 | 4 | 584 | ||
| AK308002.1 | 5 | 742 | ||
| AK313745.1 | 5 | 690 | ||
| BC058923.1 | 5 | 694 | AAH58923.1 | |
| BJ989518.1 | ||||
| BX647998.1 | 5 | 709 | ||
| M29580.1 | 5 | 851 | AAA61313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023752 | Chr.8:144835600 - 144866312 on Build GRCh38 | Gain |
|
| nsv522459 | Chr.8:144519234 - 144845293 on Build GRCh38 | Loss |
|
| nsv1031737 | Chr.8:144830982 - 144986209 on Build GRCh38 | Gain |
|
| nsv612969 | Chr.8:144785726 - 144877525 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map