Assay Details
Target Gene Details
Entrez Gene ID: | 166336 |
Gene Name: | prickle planar cell polarity protein 2 |
Gene Aliases: |
EPM5 |
Location: |
Chr.3:64093850-64445476 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE2 | NM_198859.3 | 8 | 2824 | NP_942559.1 |
| XM_011533432.2 | 10 | 2708 | XP_011531734.1 | |
| XM_011533433.2 | 9 | 2618 | XP_011531735.1 | |
| XM_011533434.2 | 10 | 2533 | XP_011531736.1 | |
| XM_011533435.2 | 9 | 2443 | XP_011531737.1 | |
| XM_011533436.2 | 9 | 2766 | XP_011531738.1 | |
| XM_011533437.2 | 9 | 11836 | XP_011531739.1 | |
| XM_011533438.2 | 6 | 2658 | XP_011531740.1 | |
| XM_011533440.2 | 10 | 2638 | XP_011531742.1 | |
| XM_017005798.1 | 8 | 2548 | XP_016861287.1 | |
| XM_017005799.1 | 7 | 4297 | XP_016861288.1 | |
| AK127113.1 | 1 | 839 | ||
| AK127839.1 | ||||
| AL833539.1 | 8 | 2825 | ||
| BC119002.1 | 8 | 2805 | ||
| BX537915.1 | 2 | 652 | CAD97898.1 | |
| BX647797.1 | 3 | 1521 | ||
| CR936678.1 | 2 | 874 |
Target Gene Details
Entrez Gene ID: | 100652759 |
Gene Name: | PRICKLE2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:64067964-64103131 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE2-AS1 | NR_045697.1 | 3 | 2722 | |
| AL832189.1 | 3 | 2722 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834717 | Chr.3:64086900 - 64265228 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map