Assay Details
Target Gene Details
Entrez Gene ID: | 116985 |
Gene Name: | ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1 |
Gene Aliases: |
CENTD2 |
Location: |
Chr.11:72685069-72752389 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 33 - Intron 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARAP1 | NM_001040118.2 | NP_001035207.1 | ||
| NM_001135190.1 | NP_001128662.1 | |||
| NM_015242.4 | NP_056057.2 | |||
| AB018325.2 | ||||
| AF411983.1 | AAL04167.1 | |||
| AJ621557.1 | CAF21317.1 | |||
| AJ706973.1 | ||||
| AK160383.1 | BAD18726.1 | |||
| AY049732.1 | AAL12169.1 | |||
| AY553630.1 | ||||
| BC008315.1 | AAH08315.1 | |||
| BC056401.1 | AAH56401.1 | |||
| BC140792.1 | ||||
| BC144562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523601 | Chr.11:72570840 - 72719890 on Build GRCh38 | Loss |
|
| nsv832207 | Chr.11:72561706 - 72758077 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map