Assay Details
Target Gene Details
Entrez Gene ID: | 10125 |
Gene Name: | RAS guanyl releasing protein 1 |
Gene Aliases: |
CALDAG-GEFI, CALDAG-GEFII, RASGRP, V, hRasGRP1 |
Location: |
Chr.15:38488097-38564806 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASGRP1 | NM_001128602.1 | NP_001122074.1 | ||
| NM_001306086.1 | NP_001293015.1 | |||
| NM_005739.3 | NP_005730.2 | |||
| XM_005254114.2 | XP_005254171.1 | |||
| XM_011521151.2 | XP_011519453.1 | |||
| XM_017021860.1 | XP_016877349.1 | |||
| AB208848.1 | BAD92085.1 | |||
| AF081195.1 | AAC79699.1 | |||
| AF081197.1 | AAF21898.1 | |||
| AF106071.1 | AAC97349.1 | |||
| AK295097.1 | ||||
| AK314091.1 | ||||
| AK316067.1 | ||||
| AY634315.2 | ||||
| AY858556.2 | ||||
| AY954625.4 | ||||
| AY966005.1 | AAX76907.1 | |||
| BC067298.1 | AAH67298.1 | |||
| BC109296.1 | AAI09297.1 | |||
| BC109297.1 | AAI09298.1 |
More Information
Set Membership: |
|

Genomic Map