Assay Details
Target Gene Details
Entrez Gene ID: | 2774 |
Gene Name: | G protein subunit alpha L |
Gene Aliases: |
DYT25 |
Location: |
Chr.18:11688956-11885685 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAL | NM_182978.3 | 1 | 756 | NP_892023.1 |
| XM_006722324.2 | 1 | 815 | XP_006722387.1 | |
| AK090868.1 | 1 | 194 | BAC03535.1 | |
| AK295088.1 | 2 | 166 | ||
| AK307260.1 | 1 | 543 | ||
| BC050021.1 | 1 | 507 | AAH50021.1 | |
| BP230966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
| nsv518207 | Chr.18:11670846 - 11695677 on Build GRCh38 | Loss |
|
| nsv516128 | Chr.18:11683432 - 11695677 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map