Assay Details
Target Gene Details
Entrez Gene ID: | 10212 |
Gene Name: | DEAD-box helicase 39A |
Gene Aliases: |
BAT1, BAT1L, DDX39, DDXL, URH49 |
Location: |
Chr.19:14408798-14419383 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX39A | NM_005804.3 | NP_005795.2 | ||
| NR_046366.2 | ||||
| XM_006722606.3 | XP_006722669.1 | |||
| XM_011527620.1 | XP_011525922.1 | |||
| XM_011527621.2 | XP_011525923.1 | |||
| AK301847.1 | ||||
| AL512698.1 | 2 | 1407 | CAH10601.1 | |
| BC001009.2 | AAH01009.1 | |||
| BC032128.2 | AAH32128.1 | |||
| BM792110.1 | ||||
| U90426.1 | AAB50231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160590 | Chr.19:14398750 - 14462842 on Build GRCh38 | Deletion |
|
| nsv833764 | Chr.19:14381787 - 14504061 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map