Assay Details
Target Gene Details
Entrez Gene ID: | 57332 |
Gene Name: | chromobox 8 |
Gene Aliases: |
PC3, RC1 |
Location: |
Chr.17:79794377-79797116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBX8 | NM_020649.2 | NP_065700.1 | ||
| AF174482.1 | AAG09180.1 | |||
| AF266479.2 | AAF76328.2 | |||
| AK074560.1 | BAC11061.1 | |||
| BC008937.2 | AAH08937.1 | |||
| BC009376.2 | AAH09376.1 | |||
| BC019289.1 | AAH19289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv538n67 | Chr.17:79765152 - 79848648 on Build GRCh38 | Gain |
|
| nsv469865 | Chr.17:79770348 - 79957778 on Build GRCh38 | Loss |
|
| nsv828116 | Chr.17:79748992 - 79855333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map