Assay Details
Target Gene Details
Entrez Gene ID: | 9388 |
Gene Name: | lipase G, endothelial type |
Gene Aliases: |
EDL, EL, PRO719 |
Location: |
Chr.18:49560699-49592908 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPG | NM_001308006.1 | NP_001294935.1 | ||
| NM_006033.3 | 5 | 973 | NP_006024.1 | |
| XM_005258390.1 | 5 | 1790 | XP_005258447.1 | |
| XM_011526265.2 | XP_011524567.1 | |||
| XM_011526267.1 | 5 | 1360 | XP_011524569.1 | |
| XM_017026095.1 | 5 | 1472 | XP_016881584.1 | |
| AF118767.1 | 5 | 947 | AAD30434.1 | |
| AK125344.1 | 5 | 1790 | ||
| AK291799.1 | 5 | 947 | ||
| AK300333.1 | ||||
| AK315252.1 | 5 | 975 | ||
| AY358928.1 | 5 | 917 | AAQ89287.1 | |
| BC060825.1 | 5 | 944 | AAH60825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map