Assay Details
Target Gene Details
Entrez Gene ID: | 23235 |
Gene Name: | salt inducible kinase 2 |
Gene Aliases: |
LOH11CR1I, QIK, SIK-2, SNF1LK2 |
Location: |
Chr.11:111602391-111726917 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIK2 | NM_015191.2 | 1 | 246 | NP_056006.1 |
| AB018324.3 | 1 | 147 | ||
| AB084424.1 | 1 | 67 | BAB91442.1 | |
| AB096248.1 | 1 | 152 | ||
| AK000396.1 | 1 | 152 | ||
| AK291233.1 | 1 | 181 | ||
| AL136764.2 | 1 | 169 | CAB66698.1 | |
| BC113459.1 | 1 | 103 | ||
| BC117183.1 | 1 | 103 | ||
| BM799630.1 | 1 | 200 | ||
| DA333935.1 | 1 | 246 |
More Information
Set Membership: |
|

Genomic Map