Assay Details
Target Gene Details
Entrez Gene ID: | 5378 |
Gene Name: | PMS1 homolog 1, mismatch repair system component |
Gene Aliases: |
HNPCC3, MLH2, PMSL1, hPMS1 |
Location: |
Chr.2:189784085-189877629 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
More Information
Set Membership: |
|

Genomic Map