Assay Details
Target Gene Details
Entrez Gene ID: | 55252 |
Gene Name: | additional sex combs like 2, transcriptional regulator |
Gene Aliases: |
ASXH2 |
Location: |
Chr.2:25733758-25878485 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASXL2 | NM_018263.4 | 13 | 5217 | NP_060733.4 |
| XM_006712039.3 | 9 | 7338 | XP_006712102.1 | |
| XM_006712040.2 | 9 | 4624 | XP_006712103.1 | |
| XM_011532950.2 | 12 | 5256 | XP_011531252.1 | |
| XM_011532951.2 | 11 | 4944 | XP_011531253.1 | |
| XM_017004429.1 | 13 | 5047 | XP_016859918.1 | |
| XM_017004430.1 | 10 | 7404 | XP_016859919.1 | |
| AB051472.2 | 13 | 5217 | ||
| AB084279.1 | 13 | 5080 | BAD00086.1 | |
| AK001760.1 | 1 | 2330 | BAA91889.1 | |
| BX647527.1 | 14 | 5290 | ||
| BX648218.1 | 13 | 5242 | ||
| CR933613.1 | 13 | 5261 | CAI45930.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map