Assay Details
Target Gene Details
Entrez Gene ID: | 5520 |
Gene Name: | protein phosphatase 2 regulatory subunit Balpha |
Gene Aliases: |
B55A, B55ALPHA, PR52A, PR55A |
Location: |
Chr.8:26291491-26372680 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R2A | NM_002717.3 | NP_002708.1 | ||
| XM_005273559.1 | XP_005273616.1 | |||
| XM_005273560.1 | XP_005273617.1 | |||
| XM_011544567.2 | XP_011542869.1 | |||
| AK314823.1 | ||||
| AY189686.1 | AAO85219.1 | |||
| BC041071.1 | AAH41071.1 | |||
| BX440554.2 | ||||
| DB451813.1 | ||||
| M64929.1 | AAA36490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1076075 | Chr.8:26291483 - 26292484 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map