Assay Details
Target Gene Details
Entrez Gene ID: | 8697 |
Gene Name: | cell division cycle 23 |
Gene Aliases: |
ANAPC8, APC8, CUT23 |
Location: |
Chr.5:138187648-138213343 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC23 | NM_004661.3 | 16 | 2256 | NP_004652.2 |
| AF053977.1 | 16 | 2223 | AAC70920.1 | |
| BC017713.1 | 16 | 2215 | AAH17713.1 |
More Information
Set Membership: |
|

Genomic Map