Assay Details
Target Gene Details
Entrez Gene ID: | 202374 |
Gene Name: | serine/threonine kinase 32A |
Gene Aliases: |
YANK1 |
Location: |
Chr.5:147234963-147401996 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK32A | NM_001112724.1 | 2 | 236 | NP_001106195.1 |
| NM_001287740.1 | 2 | 236 | NP_001274669.1 | |
| NM_145001.3 | 2 | 236 | NP_659438.1 | |
| XM_011537577.2 | 2 | 289 | XP_011535879.1 | |
| XM_011537579.2 | 2 | 289 | XP_011535881.1 | |
| XM_017009213.1 | 2 | 289 | XP_016864702.1 | |
| AK094580.1 | 2 | 289 | ||
| AK299099.1 | 2 | 127 | ||
| AK315942.1 | 2 | 226 | ||
| BC021666.1 | 2 | 149 | AAH21666.1 | |
| DA110948.1 | 2 | 236 | ||
| DR006392.1 | 2 | 128 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599938 | Chr.5:146789948 - 147426802 on Build GRCh38 | Gain |
|
| nsv519847 | Chr.5:146786815 - 147433084 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map