Assay Details
Target Gene Details
Entrez Gene ID: | 10659 |
Gene Name: | CUGBP, Elav-like family member 2 |
Gene Aliases: |
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR |
Location: |
Chr.10:10462515-11336675 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
Target Gene Details
Entrez Gene ID: | 414196 |
Gene Name: | CELF2 antisense RNA 1 |
Gene Aliases: |
C10orf31 |
Location: |
Chr.10:11316833-11344674 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF2-AS1 | NR_126062.1 | |||
| DB072070.1 | ||||
| HY033067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732940 | Chr.10:11332387 - 11332856 on Build GRCh38 | Deletion |
|
| esv2043576 | Chr.10:11332430 - 11332843 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map