Assay Details
Target Gene Details
Entrez Gene ID: | 1825 |
Gene Name: | desmocollin 3 |
Gene Aliases: |
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP |
Location: |
Chr.18:30989365-31042815 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSC3 | NM_001941.4 | 16 | 3707 | NP_001932.2 |
| NM_024423.3 | 17 | 3750 | NP_077741.2 | |
| AF297090.1 | 5 | 1814 | AAG17896.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833613 | Chr.18:30866488 - 31043633 on Build GRCh38 | Gain+Loss |
|
| nsv470414 | Chr.18:30986699 - 30996567 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map