Assay Details
Target Gene Details
Entrez Gene ID: | 767558 |
Gene Name: | leucine zipper protein 6 |
Gene Aliases: |
MPD6, MTPNUT |
Location: |
Chr.7:135926755-135977456 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP6 | NM_001128619.2 | 4 | 2218 | NP_001122091.2 |
| AA742560.1 |
Target Gene Details
Entrez Gene ID: | 136319 |
Gene Name: | myotrophin |
Gene Aliases: |
GCDP, V-1 |
Location: |
Chr.7:135926755-135977456 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTPN | NM_145808.3 | 4 | 2218 | NP_665807.1 |
| AK055660.1 | 1 | 769 | ||
| AK056225.1 | ||||
| AL834231.1 | 4 | 2090 | CAD38909.1 |
More Information
Set Membership: |
|

Genomic Map