Assay Details
Target Gene Details
Entrez Gene ID: | 9516 |
Gene Name: | lipopolysaccharide induced TNF factor |
Gene Aliases: |
PIG7, SIMPLE, TP53I7 |
Location: |
Chr.16:11547722-11636377 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LITAF | NM_001136472.1 | 4 | 1927 | NP_001129944.1 |
| NM_001136473.1 | 5 | 2025 | NP_001129945.1 | |
| NM_004862.3 | 4 | 2090 | NP_004853.2 | |
| NR_024320.1 | 4 | 1991 | ||
| XM_006720982.2 | 4 | 2001 | XP_006721045.1 | |
| XM_006720983.3 | 4 | 2291 | XP_006721046.1 | |
| XM_006720984.3 | 4 | 2043 | XP_006721047.1 | |
| XM_006720985.3 | 4 | 2005 | XP_006721048.1 | |
| XM_011522754.2 | 6 | 2404 | XP_011521056.1 | |
| XM_017023896.1 | 4 | 1950 | XP_016879385.1 | |
| AB034747.1 | 4 | 2090 | BAB32547.1 | |
| AK095955.1 | 5 | 2025 | ||
| BX537543.1 | 4 | 1992 | CAD97778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2729n100 | Chr.16:11483511 - 11548386 on Build GRCh38 | Gain |
|
| nsv524676 | Chr.16:11475598 - 11701723 on Build GRCh38 | Loss |
|
| nsv457409 | Chr.16:11449038 - 11728210 on Build GRCh38 | Gain |
|
| nsv1160341 | Chr.16:11519313 - 11553636 on Build GRCh38 | Deletion |
|
| esv3582010 | Chr.16:11543708 - 11554324 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map