Assay Details
Target Gene Details
Entrez Gene ID: | 2850 |
Gene Name: | G protein-coupled receptor 27 |
Gene Aliases: |
SREB1 |
Location: |
Chr.3:71754050-71755177 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR27 | NM_018971.1 | 1 | 1034 | NP_061844.1 |
| AB040799.1 | 1 | 1034 | BAA96645.1 | |
| HQ709187.1 | 1 | 1034 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590600 | Chr.3:71717301 - 71780868 on Build GRCh38 | Loss |
|
| esv28633 | Chr.3:71753024 - 71755305 on Build GRCh38 | Loss |
|
| nsv524847 | Chr.3:71745256 - 71772339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map