Assay Details
Target Gene Details
Entrez Gene ID: | 200895 |
Gene Name: | dihydrofolate reductase 2 |
Gene Aliases: |
DHFRL1, DHFRP4 |
Location: |
Chr.3:94057922-94063656 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHFR2 | NM_001195643.1 | 2 | 3966 | NP_001182572.1 |
| NM_176815.4 | 2 | 3853 | NP_789785.1 | |
| XM_011512537.2 | 2 | 4031 | XP_011510839.1 | |
| AW104373.1 | 1 | 82 | ||
| BC045541.1 | 2 | 3684 | AAH45541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590987 | Chr.3:93989976 - 94060820 on Build GRCh38 | Loss |
|
| esv3893746 | Chr.3:93792469 - 94164796 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145016958] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map