Assay Details
Target Gene Details
Entrez Gene ID: | 2254 |
Gene Name: | fibroblast growth factor 9 |
Gene Aliases: |
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3 |
Location: |
Chr.13:21671076-21704501 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF9 | NM_002010.2 | 1 | 912 | NP_002001.1 |
| AK290792.1 | 1 | 912 | ||
| BC069692.1 | 1 | 204 | AAH69692.1 | |
| BC103978.1 | 1 | 98 | AAI03979.1 | |
| BC103979.1 | 1 | 98 | AAI03980.1 | |
| D14838.1 | 1 | 252 | BAA03572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832560 | Chr.13:21503583 - 21692052 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map