Assay Details
Target Gene Details
Entrez Gene ID: | 253982 |
Gene Name: | aspartate beta-hydroxylase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.16:29900392-29919860 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPHD1 | AF070642.1 | |||
| BC126319.1 | ||||
| BC126321.1 | ||||
| BC144152.1 |
Target Gene Details
Entrez Gene ID: | 253980 |
Gene Name: | potassium channel tetramerization domain containing 13 |
Gene Aliases: |
BACURD1, FKSG86, PDIP1, POLDIP1, hBACURD1 |
Location: |
Chr.16:29906335-29926247 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD13 | NM_178863.4 | NP_849194.1 | ||
| NR_110933.1 | ||||
| XM_011545783.2 | XP_011544085.1 | |||
| XM_011545784.2 | XP_011544086.1 | |||
| XM_017023104.1 | XP_016878593.1 | |||
| AF289573.1 | AAL55757.1 | |||
| AF401315.2 | ||||
| AK097407.1 | ||||
| AK289630.1 | ||||
| AY027918.1 | AAK27301.1 | |||
| BC036228.1 | AAH36228.1 |
More Information
Set Membership: |
|

Genomic Map