Assay Details
Target Gene Details
Entrez Gene ID: | 2549 |
Gene Name: | GRB2 associated binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.4:143336830-143474565 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAB1 | NM_002039.3 | 1 | 263 | NP_002030.2 |
| NM_207123.2 | 1 | 263 | NP_997006.1 | |
| AK074381.1 | 1 | 726 | BAB85065.1 | |
| AK289767.1 | 1 | 206 | ||
| BC064848.1 | 1 | 127 | AAH64848.1 | |
| DA109618.1 | 1 | 331 | ||
| U43885.1 | AAC50380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026110 | Chr.4:143318672 - 143370995 on Build GRCh38 | Gain |
|
| nsv1033902 | Chr.4:143193260 - 143364619 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map