Assay Details
Target Gene Details
Entrez Gene ID: | 10207 |
Gene Name: | PATJ, crumbs cell polarity complex component |
Gene Aliases: |
Cipp, INADL, InaD-like, hINADL |
Location: |
Chr.1:61742477-62163920 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PATJ | NM_176877.2 | 4 | 361 | NP_795352.2 |
XM_005270341.3 | 4 | 359 | XP_005270398.1 | |
XM_005270347.1 | 4 | 359 | XP_005270404.1 | |
XM_006710278.3 | 4 | 359 | XP_006710341.1 | |
XM_011540462.2 | 4 | 359 | XP_011538764.1 | |
XM_011540463.2 | 4 | 547 | XP_011538765.1 | |
XM_011540464.2 | 4 | 359 | XP_011538766.1 | |
XM_011540465.2 | 4 | 359 | XP_011538767.1 | |
XM_011540466.2 | 4 | 359 | XP_011538768.1 | |
XM_011540467.2 | 4 | 359 | XP_011538769.1 | |
XM_011540468.2 | 4 | 359 | XP_011538770.1 | |
XM_011540469.2 | 4 | 359 | XP_011538771.1 | |
XM_011540470.1 | 4 | 359 | XP_011538772.1 | |
XM_016999998.1 | 4 | 359 | XP_016855487.1 | |
XM_016999999.1 | 4 | 359 | XP_016855488.1 | |
AF397170.1 | 3 | 247 | AAM28433.1 | |
AJ001306.1 | 4 | 361 | CAA04666.1 | |
AJ224747.1 | 4 | 361 | CAA12112.1 | |
AJ224748.1 | 4 | 361 | CAA12113.1 | |
AK307713.1 | 4 | 358 | ||
AK308428.1 | 4 | 358 | ||
BC142661.1 | 4 | 344 | ||
BC142723.1 | 4 | 344 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
