Assay Details
Target Gene Details
Entrez Gene ID: | 6271 |
Gene Name: | S100 calcium binding protein A1 |
Gene Aliases: |
S100, S100-alpha, S100A |
Location: |
Chr.1:153628397-153632037 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A1 | NM_006271.1 | NP_006262.1 | ||
| AB451316.1 | ||||
| AB451446.1 | ||||
| AK126904.1 | ||||
| AK312152.1 | ||||
| BC014392.1 | AAH14392.1 | |||
| BT006938.1 | AAP35584.1 | |||
| KU178317.1 | ||||
| KU178318.1 | ||||
| X58079.1 | CAA41107.1 |
Target Gene Details
Entrez Gene ID: | 6284 |
Gene Name: | S100 calcium binding protein A13 |
Gene Aliases: |
- |
Location: |
Chr.1:153618799-153634092 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A13 | NM_001024210.1 | NP_001019381.1 | ||
| XM_005245434.3 | XP_005245491.1 | |||
| XM_011509862.2 | XP_011508164.1 | |||
| XM_011509863.2 | XP_011508165.1 | |||
| XM_011509864.1 | XP_011508166.1 | |||
| XM_017002034.1 | XP_016857523.1 | |||
| XM_017002035.1 | XP_016857524.1 | |||
| AK097132.1 | ||||
| CK817939.1 |
More Information
Set Membership: |
|

Genomic Map