Assay Details
Target Gene Details
Entrez Gene ID: | 6536 |
Gene Name: | solute carrier family 6 member 9 |
Gene Aliases: |
GLYT1 |
Location: |
Chr.1:43996483-44031492 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A9 | NM_001024845.2 | 5 | 772 | NP_001020016.1 |
| NM_001261380.1 | 4 | 719 | NP_001248309.1 | |
| NM_006934.3 | 4 | 764 | NP_008865.2 | |
| NM_201649.3 | 5 | 926 | NP_964012.2 | |
| NR_048548.1 | 4 | 764 | ||
| XM_011542017.2 | 5 | 745 | XP_011540319.1 | |
| XM_017002151.1 | 5 | 680 | XP_016857640.1 | |
| XM_017002152.1 | 4 | 654 | XP_016857641.1 | |
| XM_017002153.1 | 4 | 425 | XP_016857642.1 | |
| AI884725.1 | ||||
| AK296438.1 | 4 | 522 | ||
| AK298611.1 | 3 | 377 | ||
| AK304057.1 | 5 | 640 | ||
| AK311136.1 | 5 | 635 | ||
| AK315233.1 | 4 | 767 | ||
| AK315933.1 | 4 | 585 | ||
| S70609.1 | 5 | 910 | AAB30784.1 | |
| S70612.1 | 4 | 748 | AAB30785.1 |
More Information
Set Membership: |
|

Genomic Map