Assay Details
Target Gene Details
Entrez Gene ID: | 51174 |
Gene Name: | tubulin delta 1 |
Gene Aliases: |
TUBD |
Location: |
Chr.17:59859467-59892948 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBD1 | NM_001193609.1 | 8 | 2100 | NP_001180538.1 |
| NM_001193610.1 | 8 | 2094 | NP_001180539.1 | |
| NM_001193611.1 | 7 | 1959 | NP_001180540.1 | |
| NM_001193612.1 | 9 | 2196 | NP_001180541.1 | |
| NM_001193613.1 | 6 | 1689 | NP_001180542.1 | |
| NM_016261.3 | 9 | 2265 | NP_057345.2 | |
| XM_005257425.1 | 8 | 2044 | XP_005257482.1 | |
| XM_011524871.2 | 8 | 2122 | XP_011523173.1 | |
| XM_017024716.1 | 8 | 2079 | XP_016880205.1 | |
| XM_017024717.1 | 7 | 1910 | XP_016880206.1 | |
| XM_017024718.1 | 6 | 1699 | XP_016880207.1 | |
| XM_017024719.1 | 7 | 1886 | XP_016880208.1 | |
| XM_017024720.1 | 6 | 1675 | XP_016880209.1 | |
| AK024096.1 | 5 | 1275 | BAB14825.1 | |
| AK024741.1 | 6 | 1525 | ||
| BX119803.1 | 1 | 259 |
More Information
Set Membership: |
|

Genomic Map