Assay Details
Target Gene Details
Entrez Gene ID: | 1534 |
Gene Name: | cytochrome b561 |
Gene Aliases: |
CYB561A1, FRRS2 |
Location: |
Chr.17:63432304-63446361 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB561 | NM_001017916.1 | 6 | 1320 | NP_001017916.1 |
| NM_001017917.1 | 6 | 1153 | NP_001017917.1 | |
| NM_001915.3 | 6 | 1150 | NP_001906.3 | |
| XM_005257091.1 | 6 | 1095 | XP_005257148.1 | |
| XM_017024260.1 | 6 | 1192 | XP_016879749.1 | |
| XM_017024261.1 | 5 | 1059 | XP_016879750.1 | |
| XM_017024262.1 | 6 | 1385 | XP_016879751.1 | |
| AK095244.1 | 6 | 1367 | ||
| AK294630.1 | ||||
| AK301541.1 | 5 | 922 | ||
| AK303177.1 | 6 | 1019 | ||
| BC000021.1 | AAH00021.1 | |||
| BC002976.1 | 6 | 1107 | AAH02976.1 | |
| BC091485.1 | 6 | 1121 | AAH91485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064059 | Chr.17:63271412 - 63453079 on Build GRCh38 | Gain |
|
| nsv952354 | Chr.17:63432740 - 63462739 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map