Assay Details
Target Gene Details
Entrez Gene ID: | 55316 |
Gene Name: | radical S-adenosyl methionine domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:50478800-50485980 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSAD1 | NM_018346.2 | 4 | 745 | NP_060816.1 |
| NR_130911.1 | 2 | 406 | ||
| AK002026.1 | 4 | 716 | BAA92043.1 | |
| AK022105.1 | 4 | 685 | BAB13962.1 | |
| AK222472.1 | 4 | 696 | BAD96192.1 | |
| AK297655.1 | 2 | 346 | ||
| BC005854.2 | 3 | 497 | AAH05854.4 | |
| BC050538.1 | 4 | 676 | AAH50538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2081 | Chr.17:50455308 - 50499820 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map