Assay Details
Target Gene Details
Entrez Gene ID: | 9700 |
Gene Name: | extra spindle pole bodies like 1, separase |
Gene Aliases: |
ESP1, SEPA |
Location: |
Chr.12:53268257-53293643 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESPL1 | NM_012291.4 | 3 | 407 | NP_036423.4 |
| XM_006719705.3 | 3 | 474 | XP_006719768.1 | |
| XM_011539024.2 | 3 | 435 | XP_011537326.1 | |
| XM_011539025.2 | 3 | 446 | XP_011537327.1 | |
| XM_017020253.1 | 3 | 525 | XP_016875742.1 | |
| AY455930.1 | 2 | 316 | AAR18247.1 | |
| BP244662.1 | 3 | 407 | ||
| D79987.1 | 3 | 454 | BAA11482.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss |
|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
| nsv558996 | Chr.12:53211761 - 53289202 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map