Assay Details
Target Gene Details
Entrez Gene ID: | 286183 |
Gene Name: | Na+/K+ transporting ATPase interacting 3 |
Gene Aliases: |
FAM77D |
Location: |
Chr.8:62248942-62999652 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAIN3 | NR_130764.1 | |||
| XM_017013359.1 | XP_016868848.1 | |||
| AK096949.1 | BAC04909.1 |
Target Gene Details
Entrez Gene ID: | 643763 |
Gene Name: | uncharacterized LOC643763 |
Gene Aliases: |
- |
Location: |
Chr.8:62977861-62984901 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UG0898H09 | NR_027378.1 | 1 | 3306 | |
| AF360117.1 | 1 | 3306 | AAN76520.1 | |
| AK096517.1 | 1 | 2660 | BAC04808.1 | |
| AK127279.1 | 1 | 3221 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018879 | Chr.8:62369165 - 63025073 on Build GRCh38 | Gain |
|
| nsv6227 | Chr.8:62979528 - 63024393 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map