Assay Details
Target Gene Details
Entrez Gene ID: | 81031 |
Gene Name: | solute carrier family 2 member 10 |
Gene Aliases: |
ATS, GLUT10 |
Location: |
Chr.20:46708358-46736347 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A10 | NM_030777.3 | 2 | 1129 | NP_110404.1 |
| XM_011529060.2 | 2 | 1212 | XP_011527362.1 | |
| XM_011529061.2 | 2 | 2769 | XP_011527363.1 | |
| XM_011529062.2 | 2 | 1223 | XP_011527364.1 | |
| XM_011529063.2 | 2 | 1222 | XP_011527365.1 | |
| XM_011529064.2 | 2 | 1224 | XP_011527366.1 | |
| XM_011529065.2 | 2 | 1219 | XP_011527367.1 | |
| XM_017028087.1 | 2 | 1122 | XP_016883576.1 | |
| AF248053.1 | 2 | 1282 | AAK31911.1 | |
| AF321240.1 | 2 | 1084 | AAK26294.1 | |
| AK055548.1 | 2 | 1129 | ||
| AK290961.1 | 2 | 1120 | ||
| AK315269.1 | 2 | 1120 | ||
| AL137188.3 | 1 | 1040 | CAB69822.2 | |
| BC101657.1 | 2 | 1128 | AAI01658.1 | |
| BC113423.1 | 2 | 1128 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map