Assay Details
Target Gene Details
Entrez Gene ID: | 151188 |
Gene Name: | ADP ribosylation factor like GTPase 6 interacting protein 6 |
Gene Aliases: |
AIP-6, AIP6, PFAAP1 |
Location: |
Chr.2:152717893-152761253 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL6IP6 | NM_152522.5 | NP_689735.1 | ||
| NR_024526.2 | ||||
| XM_006712326.3 | XP_006712389.1 | |||
| AF530059.1 | AAQ09943.1 | |||
| AK023109.1 | ||||
| AK094696.1 | ||||
| AK315657.1 | ||||
| AW967583.1 | ||||
| BC028741.2 | AAH28741.1 | |||
| BC070140.1 | AAH70140.1 | |||
| BP200063.1 | ||||
| BX437990.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834421 | Chr.2:152613595 - 152807721 on Build GRCh38 | Loss |
|
| esv34659 | Chr.2:150858366 - 152860232 on Build GRCh38 | Loss |
|
| esv3592922 | Chr.2:151571476 - 153829714 on Build GRCh38 | Loss |
|

Genomic Map