Assay Details
Target Gene Details
Entrez Gene ID: | 389090 |
Gene Name: | olfactory receptor family 6 subfamily B member 2 |
Gene Aliases: |
OR2-1, OR6B2P |
Location: |
Chr.2:240029491-240030429 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6B2 | NM_001005853.1 | NP_001005853.1 | ||
| BC137561.1 | 1 | 989 | ||
| BC137562.1 | 1 | 989 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|
| nsv834584 | Chr.2:239892475 - 240037701 on Build GRCh38 | Gain |
|
| nsv525278 | Chr.2:240001219 - 240122324 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map