Assay Details
Target Gene Details
Entrez Gene ID: | 56937 |
Gene Name: | prostate transmembrane protein, androgen induced 1 |
Gene Aliases: |
STAG1, TMEPAI |
Location: |
Chr.20:57648392-57711536 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMEPA1 | NM_001255976.1 | 4 | 4034 | NP_001242905.1 |
| NM_020182.4 | 4 | 4305 | NP_064567.2 | |
| NM_199169.2 | 4 | 3958 | NP_954638.1 | |
| NM_199170.2 | 4 | 3951 | NP_954639.1 | |
| NM_199171.2 | 4 | 4016 | NP_954640.1 | |
| AF305616.1 | 4 | 4232 | AAL16781.1 | |
| AJ308020.1 | ||||
| AK024618.1 | 1 | 1180 | ||
| BQ008707.1 | 1 | 630 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758802 | Chr.20:57595722 - 58113215 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map